Learn More
Thermo Scientific™ Whole Cell Stains
Provide excellent staining for high-content screening assays and other fluorescent cellular imaging methods involving fluorescence microscopy.
Manufacturer:  Thermo Scientific™ 8403201
 View more versions of this product
Catalog No. 84-032-01
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Provide excellent staining for high-content screening assays and other fluorescent cellular imaging methods involving fluorescence microscopy using Thermo Scientific™ Whole Cell Stains.
These stains are intense, highly photostable and match the output wavelengths of common fluorescence instrumentation. Effective image analysis in cell-based assays and fluorescence microscopy of the localization of specific targets requires fluorescent labeling of the entire cells, nuclei or major cellular components. Whole cell stains identify cell numbers, cell borders and cell morphologies as useful primary objects for assessment of specific protein targets and their redistribution in response to treatment conditions.
  • Provide excellent staining for HCA and fluorescence microscopy
  • Intense, highly photostable and match the output wavelengths of common fluorescence instrumentation
  • When the whole cell is the primary object used to identify and count individual cells, effectively distinguish intact cells from bordering cells (with analysis by appropriate software)
Includes:
Fluorescent stains to detect whole cells (blue, green, orange or red)
Requires:
Fluorescence microscope, Thermo Scientific™ Cellomics™ ArrayScan™ VTI or other HCS reader
Description & Specifications
Specifications
Cellomics Whole Cell Stain Green
1 × 96 tests
Whole cell (green); Ex/Em = 493/518nm