Thermo Scientific™ Whole Cell Stains

Provide excellent staining for high-content screening assays and other fluorescent cellular imaging methods involving fluorescence microscopy.

Manufacturer: Thermo Scientific™ 8403302

 View more versions of this product

Catalog No. 8403302

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Quantity 5 × 96 tests
    Target Whole cell (orange); Ex/Em = 562/576nm
    Description Cellomics Whole Cell Stain Orange

    Provide excellent staining for high-content screening assays and other fluorescent cellular imaging methods involving fluorescence microscopy using Thermo Scientific™ Whole Cell Stains.

    These stains are intense, highly photostable and match the output wavelengths of common fluorescence instrumentation. Effective image analysis in cell-based assays and fluorescence microscopy of the localization of specific targets requires fluorescent labeling of the entire cells, nuclei or major cellular components. Whole cell stains identify cell numbers, cell borders and cell morphologies as useful primary objects for assessment of specific protein targets and their redistribution in response to treatment conditions.

    • Provide excellent staining for HCA and fluorescence microscopy
    • Intense, highly photostable and match the output wavelengths of common fluorescence instrumentation
    • When the whole cell is the primary object used to identify and count individual cells, effectively distinguish intact cells from bordering cells (with analysis by appropriate software)

    Includes:

    Fluorescent stains to detect whole cells (blue, green, orange or red)

    Requires:

    Fluorescence microscope, Thermo Scientific™ Cellomics™ ArrayScan™ VTI or other HCS reader

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit