Learn More
WHSC1L1 Mouse anti-Human, Clone: OTI6A12, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF810011
Catalog No. 50-166-9847
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is related to the Wolf-Hirschhorn syndrome candidate-1 gene and encodes a protein with PWWP (proline-tryptophan-tryptophan-proline) domains. The function of the protein has not been determined. Two alternatively spliced variants have been described.
Specifications
Specifications
WHSC1L1
Monoclonal
Unconjugated
NSD3
Lyophilized
NSD3
Human recombit protein fragment corresponding to amino acids 427-566 of human WHSC1L1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI6A12
PBS with 8% trehalose and no preservative; pH 7.3
Q9BZ95
KMT3F, KMT3G, WHISTLE, WHSC1L1, pp14328
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
54904