Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WHSC2 Mouse anti-Human, Rat, Biotin, Clone: 6B11H8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237394B

Catalog No. NBP237394B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

WHSC2
6B11H8
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
WHSC2
Purified recombinant fragment of human WHSC2 (AA: 280-511) expressed in E. Coli.
Protein A or G purified
Cancer
Primary
7469
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
FLJ10442, negative elongation factor A, NELFA, NELF-AFLJ25112, Wolf-Hirschhorn syndrome candidate 2, Wolf-Hirschhorn syndrome candidate 2 protein
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit