Learn More
WHSC2 Mouse anti-Human, Rat, DyLight 405, Clone: 6B11H8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237394V
Catalog No. NBP237394V
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
WHSC2
6B11H8
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
WHSC2
Purified recombinant fragment of human WHSC2 (AA: 280-511) expressed in E. Coli.
Protein A or G purified
Cancer
Primary
7469
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
FLJ10442, negative elongation factor A, NELFA, NELF-AFLJ25112, Wolf-Hirschhorn syndrome candidate 2, Wolf-Hirschhorn syndrome candidate 2 protein
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Rat