Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WIBG Mouse anti-Human, HRP, Clone: OTI5E2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274868H

Catalog No. NBP274868H


Add to cart

Description

Description

WIBG Monoclonal antibody specifically detects WIBG in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

WIBG
OTI5E2
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
WIBG
Full length recombinant protein of human WIBG (NP_115721) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Western Blot
HRP
PBS with No Preservative
MGC13064, partner of Y14 and mago, PYMProtein wibg homolog, within bgcn homolog (Drosophila)
Mouse
IgG1
0.1 mL
Primary
84305.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit