Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WIBG Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | WIBG |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179834
|
Novus Biologicals
NBP179834 |
100 μL |
Each of 1 for $436.00
|
|
Description
WIBG Polyclonal specifically detects WIBG in Human samples. It is validated for Western Blot.Specifications
WIBG | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
MGC13064, partner of Y14 and mago, PYMProtein wibg homolog, within bgcn homolog (Drosophila) | |
Synthetic peptide directed towards the C terminal of human WIBGThe immunogen for this antibody is WIBG. Peptide sequence KKIKNLKKKLRQVEELQQRIQAGEVSQPSKEQLEKLARRRALEEELEDLE. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
NP_115721 | |
84305 | |
IgG | |
Affinity Purified | |
23 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title