Learn More
DWK Life Sciences Wheaton™ Wide Mouth Round Bottle, HDPE, Without Cap
Wide mouth HDPE bottles with or without attached caps are available in a range of sizes, from 30mL to 3840mL.
Manufacturer:  DWK Life Sciences W218826
 View more versions of this product
Catalog No. 03-084-158
  Check Availability
Add to cart
Description
Description
  • High Density Polyethylene (HDPE) in a natural color
  • Provides good chemical resistance
  • Without caps or supplied with pre-attached white polypropylene screw caps with PE liners
Specifications
Specifications
950mL
HDPE
Wide Mouth Bottle
Wide Mouth
Natural Color
72/Case
No
Round