Learn More
WIF-1 Mouse anti-Human, Janelia Fluor 549, Clone: MM0602-6D27, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211997JF549
Catalog No. NBP21997549
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
WIF-1 Monoclonal antibody specifically detects WIF-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
WIF-1
MM0602-6D27
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
WIF1
Human WIF1
Protein G purified
Signal Transduction, Wnt Signaling Pathway
Primary
11197
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
WIF-1, WNT inhibitory factor 1
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
Human