Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WIF-1 Rat anti-Mouse, HRP, Clone: RM0144-3M51, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP122520H
Description
WIF-1 Monoclonal antibody specifically detects WIF-1 in Mouse samples. It is validated for Western Blot.Specifications
WIF-1 | |
RM0144-3M51 | |
Western Blot | |
Purified | |
WIF1 | |
Mouse recombinant protein of WIF-1 | |
Protein G purified | |
Signal Transduction, Wnt Signaling Pathway | |
Primary | |
11197 | |
Mouse |
Western Blot | |
HRP | |
PBS with No Preservative | |
WIF-1, WNT inhibitory factor 1 | |
Rat | |
IgG2 | |
0.1 ml | |
Store at 4C in the dark. | |
Monoclonal | |
This was produced from a hybridoma (mouse myeloma) fused with spleen cells from a rat immunized with mouse recombinant protein of WIF-1. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title