Learn More
Sklar InstrumentsSupplier Diversity Partner Wigmore Saw, Sklar, 7-1/2 in., Straight, Serrated, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 111530
Wigmore Saw, Sklar, 7-1/2 in., Straight, Serrated, Stainless Steel. Sklar's Wigmore cast saw has a solid handle with a serrated stainless steel blade. It is used to cut through plaster casts. The serrations offer an advantage for materials such as plaster.
Catalog No. 50-143-7320