Learn More
Wilmad™ SP Scienceware™ 18mm O.D. Medium Walled Precision NMR Tube Available on GSA/VA Contract for Federal Government customers only.
To maximize SNR, Precision NMR Tubes have minimal paramagnetic impurities that would impact shimming. Tight ID and OD tolerance as small as 0.0065mm accommodates Wilmad inserts
Manufacturer:  Wilmad™ 663521895
 View more versions of this product
Catalog No. 16-800-365
  Check Availability
Add to cart
Alert:
Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6
Specifications Description & Specifications
Specifications
228.6mm
18.000±0.013mm
17.00±0.013mm
For experiments requiring critical shimming quality (high/ultrahigh field, non-spinning, multi-dimensional, multi-nuclei, DNP experiments and studies involving biological samples)
0.50mm
51μm
25μm
350MHz
9 in.
18-P9-7-5