Learn More
Wilmad™ SP Scienceware™ 5mm O.D. Thin Walled Suprasil™ NMR Tube Available on GSA/VA Contract for Federal Government customers only.
Wilmad Precision NMR Suprasil™ (Synthetic Quartz) Tubes are manufactured from high purity synthetic fused silica materials with <0.005ppm Fe2O3 and outstanding optical characteristics in the deep UV to the near IR
Manufacturer:  Wilmad™ 665080585
 View more versions of this product
Catalog No. 16-800-462
  Check Availability
Add to cart
Alert:
Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.
Specifications Description & Specifications
Specifications
High purity synthetic fused silica materials
25μm
13μm
500MHz
8 in.
528-PP-8SUP-5
203.2mm
4.9635±0.0065mm
4.2065±0.0065mm
Ideal for photolysis experiments that employ 266nm light from a Q-Switched laser or 254nm light emitted by a mercury low pressure lamp with a Schott UG 5 filter, as well as variable temperature experiments that have a temperature step over 120°C
0.38mm