Learn More
Wilmad-LabGlass™ SP Scienceware™ 5mm O.D. Thin Walled Suprasil™ NMR Tube
Wilmad Precision NMR Suprasil™ (Synthetic Quartz) Tubes are manufactured from high purity synthetic fused silica materials with <0.005ppm Fe2O3 and outstanding optical characteristics in the deep UV to the near IR
$478.00 - $526.50
Specifications
Material High Purity Synthetic Fused Silica
View More Specs
Alert:
Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.
Includes: Tube and disposable cap
Products 12
Catalog Number Mfr. No. Model Length (Metric) Field Strength Thickness (Metric) Wall Price Quantity & Availability  
Catalog Number Mfr. No. Model Length (Metric) Field Strength Thickness (Metric) Wall Price Quantity & Availability  
16-800-340
View Documents
Wilmad-LabGlass™
663080585
507-PP-8SUP-5 203.2 mm 300MHz 0.38 mm
Pack of 5 for $490.00
Add to cart
16-800-339
View Documents
Wilmad-LabGlass™
663080575
507-PP-7SUP-5 178 mm 300MHz 0.38 mm
Pack of 5 for $478.00
Add to cart
16-800-341
View Documents
Wilmad-LabGlass™
663080595
507-PP-9SUP-5 228.6 mm 300MHz 0.38 mm
Pack of 5 for $499.50
Add to cart
16-800-461
View Documents
Wilmad-LabGlass™
665080575
528-PP-7SUP-5 178 mm 500MHz 0.38 mm
Pack of 5 for $491.00
Add to cart
16-800-463
View Documents
Wilmad-LabGlass™
665080595
528-PP-9SUP-5 228.6 mm 500MHz 0.38 mm
Pack of 5 for $517.00
Add to cart
16-800-462
View Documents
Wilmad-LabGlass™
665080585
528-PP-8SUP-5 203.2 mm 500MHz 0.38 mm
Pack of 5 for $508.00
Add to cart
16-800-486
View Documents
Wilmad-LabGlass™
666080575
535-PP-7SUP-5 178 mm 600MHz 0.38 mm
Pack of 5 for $518.00
Add to cart
16-800-487
View Documents
Wilmad-LabGlass™
666080585
535-PP-8SUP-5 203.2 mm 600MHz 0.38 mm
Pack of 5 for $526.50
Add to cart
16-800-424
View Documents
Wilmad-LabGlass™
664081075
513-7PP-7SUP-5 178 mm 400MHz 0.46 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
16-800-425
View Documents
Wilmad-LabGlass™
664081085
513-7PP-8SUP-5 203.2 mm 400MHz 0.46 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
16-800-426
View Documents
Wilmad-LabGlass™
664081095
513-7PP-9SUP-5 228.6 mm 400MHz 0.46 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
16-800-488
View Documents
Wilmad-LabGlass™
666080595
535-PP-9SUP-5 228.6 mm 600MHz 0.38 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Manufactured in a state-of-the-art ISO 9001:2008 USA facility using a unique precision shrinking and grinding process to shape the inner surface with maximized filling factor.
  • Transmission rate from 190nm to 2600nm is well over 95% (10mm thickness) excluding reflection
  • Possesses a similar thermal expansion rate and tensile strength as natural quartz which is 14 times more robust during the cooling/heating process than Type 1 Class A glass
  • Tight I.D. and O.D. tolerance as small as 0.0065mm accommodates Wilmad inserts
  • To maximize SNR, Wilmad Precision NMR Suprasil™ Tubes have minimal paramagnetic impurities that would impact shimming
  • Inner surface is resistant to strong acid and base at ambient temperature
  • Safe for experiments at temperatures up to 1300°C
  • 100% inspection with multiple NIST traceable gauges and optical surface defect checks
Specifications
Specifications
High Purity Synthetic Fused Silica