Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Wilmad™ 5mm OD Thin Walled 2 in. Raman Spectroscopy Tube

Designed specifically for Raman experiments - 100% inspected to ensure the success of your experiment.

Manufacturer:  Bel-Art™ 660000082

Catalog No. 16-800-182


Add to cart

Alert:

Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.


Description

Description

  • Made from borosilicate glass; meets ASTM E438 Type 1 Class B standard, recognized as N51A
  • 100% visual and physical inspection, including physical dimensions and surface defects, ensures quality,  
  • Manufactured in an ISO 9001:2008 USA facility
  • Cooling/heating of these tubes may lead to breakage; for experiments that involve cooling or heating, refer to Wilmad Precision Tubes
Specifications

Specifications

51 mm
4.947±0.019 mm
Raman Spectroscopy
2 in.
Wilmad™ WG-5MM-ECONOMY-2
Borosilicate Glass
High-Throughput
Tube and disposable cap
0.43 mm
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit