Learn More
Wilmad™ SP Scienceware™ 8mm O.D. Thin Walled Precision NMR Tube Available on GSA/VA Contract for Federal Government customers only.
To maximize SNR, Precision NMR Tubes have minimal paramagnetic impurities that would impact shimming. Tight I.D. and O.D. tolerance as small as 0.0065mm accommodates Wilmad inserts
Manufacturer:  Wilmad™ 663500875
 View more versions of this product
Catalog No. 16-800-345
  Check Availability
Add to cart
Alert:
Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.
Specifications Description & Specifications
Specifications
178mm
7.9935±0.0065mm
6.990±0.013mm
For experiments requiring critical shimming quality (high/ultrahigh field, non-spinning, multi-dimensional, multi-nuclei, DNP experiments and studies involving biological samples)
0.51mm
51μm
25μm
350MHz
7 in.
513A-5PP-7-5