Wilmad™ Thin Walled High Throughput NMR Tubes Available on GSA/VA Contract for Federal Government customers only.

Possess an average camber of 60 microns to guarantee the clarity of the spectrum for small molecule (MW<250) samples up to 600 MHz. Wilmad Thin Walled High Throughput NMR Tubes are designed for routine use in most low to mid field NMR spectrometers.

Manufacturer: Wilmad™ 669995700

 View more versions of this product

Catalog No. 16-800-495

  Check Availability
Add to Cart

    Specifications

    Length (Metric) 178mm
    Diameter (Metric) Outside 4.94665 ± 0.01905mm
    Camber (English) 60μm
    Length (English) 7 in.
    Thickness (Metric) Wall 0.43mm
    Quantity 100/pk
    Description MHz Rating: High-Throughput
    Closure Disposable NMR cap
    Model WG-1000-7
    Material ASTM E438 Type 1 Class B glass
    Product Type NMR Tubes

    • Camber and concentricity values listed are the Total Indicator Reading (TIR)
    • Designed for routine use in most low to mid field NMR spectrometers
    • One of the best OD tolerances in the industry
    • Guaranteed to fit tightly in spinner turbine
    • Made from ASTM E438 Type 1 Class B glass
    • 100% inspected for surface defects and physical dimension to ensure the success of your experiments
    • WG-1000-4-SJ and WG-1000-7-SJ are supplied with authentic Bruker SampleJet caps, all others come with a disposable NMR cap
    Note: For NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, please refer to Wilmad Precision NMR Tubes.

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit