Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Coaxial Small Volume NMR Tubes

Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Coaxial Small Volume NMR Tubes feature a tube within a tube design that allows for easy external referencing.

$24.75 - $56.75

Specifications

Field Strength < 600 MHz
Material Borosilicate Glass
View More Specs

Products 5
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Model Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Model Price Quantity & Availability  
16-800-103
View Documents
Bel-Art™ SP Scienceware™
660000003
4.2 mm outer tube, 2.34 mm inner tube 5 mm outer tube, 3.3 mm inner tube 517-Complete
Each for $56.75
Add to cart
 
16-800-104
View Documents
Bel-Art™ SP Scienceware™
660000004
2.34 mm 3.30 mm 517-Inner
Each for $25.25
Add to cart
 
16-800-105
View Documents
Bel-Art™ SP Scienceware™
660000005
4.2 mm outer tube, 1.96 mm inner tube 5 mm outer tube, 2.97 mm inner tube 518-Complete
Each for $56.75
Add to cart
 
16-800-106
View Documents
Bel-Art™ SP Scienceware™
660000006
4.2 mm outer tube, 1.5 mm inner tube 5 mm outer tube, 2.52 mm inner tube 519-Complete
Each for $56.75
Add to cart
 
16-800-107
View Documents
Bel-Art™ SP Scienceware™
660000007
1.50 mm 2.52 mm 519-Inner
Each for $24.75
Add to cart
 
Description

Description

  • In complete sets, the tubes are fused together at the bottom
  • Recommended for high field experiments under 600 MHz
  • Tight ID and OD tolerance as small as 0.0065 mm
  • Separately wrapped spacer should replace cap during experiments for improved stability
  • Three unique sample ratios are available for a wide range of applications
  • Complete sets come with inner and outer tubes and cushioning cap for protection during transportation
Specifications

Specifications

< 600 MHz
Borosilicate Glass
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit