Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Spherical NMR Bulb Microcell Insert
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Spherical NMR Bulb Microcell Insert is ideal for experiments that need to center small volume samples in the Rf coil such as magnetic susceptibility measurement.
Manufacturer:  Bel-Art™ SP Scienceware™ 660000008
Catalog No. 16-800-108
Add to cart
Description
Description
  • Sample volume can be as little as 18 µl
  • 3 different inserts with various shapes (Spherical Bulb, Capillary Tube and Cylindrical Bulb)
  • Outer tube (Precision Thin Walled) must be ordered separately depending on magnetic field strength.
  • Not recommended for variable temperature experiments.
  • Use our PTFE Needles (#90630) with syringes to fill and clean sample.
Specifications
Specifications
44.45 mm
Borosilicate Glass
1.5 mm
Spherical Bulb
5 mm Wilmad Precision NMR Tubes
Can contain a volume of 18 μl
1 mm
529-A
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Only use with Wilmad-LabGlass Precision Thin-Walled Tubes