Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Wilmad™ Precision Quartz EPR Sample Tube

  • Hold 10 to 15 times better tolerances on the inner diameter and 5 times smaller camber and concentricity vs. economy brand tubes
  • Ideal for continuous wave experiments that require highly reproducible sample volume in the cavity
  • Individually wrapped to prevent contamination

Supplier:  Bel-Art™ 660000016

 View more versions of this product

Catalog No. 16-800-116

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

  • Hold 10 to 15 times better tolerances on the inner diameter and 5 times smaller camber and concentricity vs. economy brand tubes
  • Ideal for continuous wave experiments that require highly reproducible sample volume in the cavity
  • Individually wrapped to prevent contamination
Specifications

Specifications

15 μm
76 μm
Sample Tube
3.20±0.013 mm
9.84 in.
715-PQ-250M
250 mm
Quartz
4.00±0.013 mm
Cap
0.40 mm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit