Learn More
Wilmad-LabGlass™ SP Scienceware™ Rotor and Caps for Bruker MAS Probe
Provide solution for analysis of solid samples
$74.00
Specifications
For Use With (Application) Solid state NMR spectrometers
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Model Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Model Price Quantity & Availability  
16-800-494
View Documents
Wilmad-LabGlass™
668090581
For Bruker MAS 3.2mm Probe 520-1A
Each for $74.00
Add to cart
16-800-491
View Documents
Wilmad-LabGlass™
668000545
For Bruker MAS 7mm Probe 528-PP-4VAR This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Manufactured from high-purity Zirconia
  • Caps fitted with O-rings for improved sealing
  • MAS Probe and caps compatible for most solid state NMR spectrometers
  • Thoroughly inspected before and after precision-machining process ensures no material irregularities
  • Spin testing performed to highest specified spinning speed, assuring performance without overspinning the rotor
Specifications
Specifications
Solid state NMR spectrometers