Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Wilmad™ Stainless Steel Micro Spatula

Fits into most 4mm or larger OD NMR/EPR tubes. Bel-Art SP Scienceware™ Wilmad™ Stainless Steel Micro Spatula make solid state and gel-phase sample transfers easier.

Manufacturer:  Bel-Art™ 660000022

Catalog No. 16-800-122


Add to cart

Description

Description

  • Fits into most 4mm or larger OD NMR/EPR tubes
  • Makes solid state and gel-phase sample transfers easier
Specifications

Specifications

Most 4mm OD or larger NMR/EPR tubes
Stainless Steel
806
250 mm
3 mm
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit