Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Superior Pressure NMR Tube Caps, 5.0mm Dia., Translucent
Ideal for experiments that require an air-tight environment under vacuum or low pressure less than 1 bar.
$7.13 - $55.80
Specifications
Diameter (Metric) Outer 5.0mm
Item Description Gas Tight - Superior Pressure
Color Translucent
Material Polyethylene
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
16-800-223 View Documents Bel-Art™
660050110
Model: Wilmad™ 521-PC-100 Pack of 100 for $7.13
Add to cart
16-800-235 View Documents Bel-Art™
660051010
Model: Wilmad™ 521-PC-1000 Pack of 1000 for $55.80
Add to cart
Description
Description
Use with medium- and heavy-walled NMR tubes for better seal and robustness in variable temperature experiments.
Specifications
Specifications
5.0mm
Gas Tight - Superior Pressure
Translucent
Polyethylene