Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Tube Washer
Wiorks with filter flask and inverted sample tube. Bel-Art SP Scienceware™ Wilmad™ Tube Washer with simple design washing unit is the most economical and reliable for single tube cleaning.
Supplier:  Bel-Art™ 660000065
 View more versions of this product
Catalog No. 16-800-165
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
  • Each unit only accommodates NMR tube with OD and length as indicated in specifications
  • Requires filter flask (sold separately; 500ml recommended)
  • Inverted sample tube is inserted into the washer and solvent is introduced into the reservoir using a series of wash bottles.
  • Pulls air through the tube to complete the procedure.
Specifications
Specifications
203 mm
Fits tube diameter: 5mm, “B” diameter: 10mm
24/40 inner joint
8 in.
Recommendations
Recommendations