WIN 64338 hydrochloride, Tocris

Bradykinin B2 antagonist

Manufacturer: R&D Systems 1057/10

Quantity 10mg

 View more versions of this product

Catalog No. 10-571-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C45H68ClN4OP·HCl
    Formula Weight 783.95
    Synonym (S)-4-[2-[Bis(cyclohexylamino)methyleneamino]-3-(2-naphthalenyl)-1-oxopropylamino]benzyl tributyl phosphonium chloride hydrochloride
    Chemical Name or Material WIN 64338 hydrochloride

    Specifications

    Formula Weight 783.95
    Quantity 10mg
    Assay Percent Range >98%
    Molecular Formula C45H68ClN4OP·HCl
    CAS 163727-74-0
    Synonym (S)-4-[2-[Bis(cyclohexylamino)methyleneamino]-3-(2-naphthalenyl)-1-oxopropylamino]benzyl tributyl phosphonium chloride hydrochloride
    Recommended Storage Desiccate at +4°C
    Chemical Name or Material WIN 64338 hydrochloride

    Desiccate at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit