Learn More
Wind Powered Generator Products for Science Education
Study the inter-relationship between aerodynamics, motion, and electrical power production.
Manufacturer:  Science FirstSupplier Diversity Partner 800
Catalog No. S435803
Add to cart
Description
Description
  • Six fully adjustable vanes describe a 36cm diameter circle
  • Vanes are coupled to the generator through a 4:1 drive train
  • DC output is monitored with a variable intensity bulb or with a voltmeter connected to the binding posts provided
Specifications
Specifications
Study the inter relationship between aerodynamics, motion, and electrical power production
36cm dia. Circle six fully adjustable vanes coupled through a 4:1 drive train