Learn More
Wing Top Accessory Products for Science Education
Creates a fan-shaped flame for glass bending.
Specifications
For Use With (Application) Create a Fan Shaped Flame for Glass Bending
Material Brass
Type Flame Spreader
View More Specs
Products 2
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) For Use With (Equipment) Price Quantity & Availability  
S49125B Products for Science Education
View Documents
Humboldt Manufacturing CompanySupplier Diversity Partner
H6670/EMD
1.06 in. 2.7 cm Burner Tube of a Bunsen Burner (S49107, S49108, S49119 And S49120) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
S49125A Products for Science Education
View Documents
Humboldt Manufacturing CompanySupplier Diversity Partner
H6650/EMD
0.51 in. 1.3 cm Burner Tube of a Bunsen Burner (S49106) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Slip this brass wing top over the burner tube of a Bunsen burner
  • 13mm burner tube fits burner S49106
  • 27mm burner tube fits burners S49107, S49108, S49119 and S49120
Specifications
Specifications
Create a Fan Shaped Flame for Glass Bending
Brass
Flame Spreader