Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WIPI2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282059F
This item is not returnable.
View return policy
Description
WIPI2 Polyclonal antibody specifically detects WIPI2 in Human, Rat samples. It is validated for ELISASpecifications
WIPI2 | |
Polyclonal | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
ELISA | |
FITC | |
ATG18B, Atg21, CGI-50, DKFZP434J154, DKFZp686P02188, FLJ12979, FLJ14217, FLJ42984, WD repeat domain phosphoinositide-interacting protein 2, WD repeat domain, phosphoinositide interacting 2, WD40 repeat protein interacting with phosphoinositides 2, WIPI-2, WIPI49-like protein 2 | |
Rabbit polyclonal WIPI2 antibody was raised against a 17 amino acid peptide near the amino terminus of human WIPI2. The immunogen is located within the first 50 amino acids of WIPI2. Amino Acid Squence: CANFNQDNTEVKGASRAA | |
0.1 mL | |
Autophagy, Cell Biology, Endocrinology | |
26100 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction