Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WIPI2 Mouse anti-Human, Mouse, Clone: 2A2, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen WIPI2
Clone 2A2
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, ELISA
Applications Flow Cytometry, ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122691
View Documents
Novus Biologicals
NBP30760910UG
100 μg
Each for $352.89
Add to cart
 
NB122692
View Documents
Novus Biologicals
NBP30760920UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

WIPI2 Monoclonal antibody specifically detects WIPI2 in Human, Mouse samples. It is validated for Flow Cytometry, ELISA
Specifications

Specifications

WIPI2
0.2 mg/ml
Flow Cytometry, ELISA
Purified
RUO
10 mM PBS with 0.05% BSA
26100
IgG1 Kappa
Primary
Store at 4°C.
2A2
Flow Cytometry 1-2 ug/million cells, ELISA
Unconjugated
Mouse
Human, Mouse
ATG18B, Atg21, CGI-50, DKFZP434J154, DKFZp686P02188, FLJ12979, FLJ14217, FLJ42984, WD repeat domain phosphoinositide-interacting protein 2, WD repeat domain, phosphoinositide interacting 2, WD40 repeat protein interacting with phosphoinositides 2, WIPI-2, WIPI49-like protein 2
Recombinant fragment (around aa 418-436) of human WIPI2 protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit