Learn More
H-B Instrument™ Wireless Weather Station
SCI_ED
Become an amateur meteorologist with these all-in-one wireless stations.
Manufacturer:  H-B Instrument™ B615000400
 View more versions of this product
Catalog No. S95624
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Batteries and remote sensor
Description
Description
  • Perpetual calendar up to year 2050 with date and day of week in seven languages (English, German, French, Italian, Spanish, Dutch, and Danish)
  • 12/24-hour selectable time with snooze alarm
  • Moon phase
  • Dual-scale indoor/outdoor temperature range — Indoor: 0°/50°C (35°/122°F); Outdoor: -20°/+60°C (-4°/+140°F)
  • Indoor/outdoor temperature trend indicator
  • RF 433MHz wireless thermometer links up to three remote sensors with 100 ft. transmission (30m)
  • Model 693 adds max./min. temperature memory and outdoor temperature alert
Order Info
Individually packaged with multi-language instructions.
Certifications
CE marked
Compliance
RoHS compliant
Specifications
Specifications
692: Weather Station Trend
Batteries and remote sensor