Learn More
H-B Instrument™ Wireless Weather Station Products for Science Education
Become an amateur meteorologist with these all-in-one wireless stations.
Specifications
Includes Batteries and remote sensor
View More Specs
Includes: Batteries and remote sensor
Products 2
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
S95625 Products for Science Education
View Documents
H-B Instrument™
B615000500
693: Weather Station Wireless This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
S95624 Products for Science Education
View Documents
H-B Instrument™
B615000400
692: Weather Station Trend This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Perpetual calendar up to year 2050 with date and day of week in seven languages (English, German, French, Italian, Spanish, Dutch, and Danish)
  • 12/24-hour selectable time with snooze alarm
  • Moon phase
  • Dual-scale indoor/outdoor temperature range — Indoor: 0°/50°C (35°/122°F); Outdoor: -20°/+60°C (-4°/+140°F)
  • Indoor/outdoor temperature trend indicator
  • RF 433MHz wireless thermometer links up to three remote sensors with 100 ft. transmission (30m)
  • Model 693 adds max./min. temperature memory and outdoor temperature alert
Order Info
Individually packaged with multi-language instructions.
Certifications
CE marked
Compliance
RoHS compliant
Specifications
Specifications
Batteries and remote sensor