Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WIZ Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318492UV
Description
WIZ Polyclonal antibody specifically detects WIZ in Human, Mouse samples. It is validated for Western BlotSpecifications
WIZ | |
Polyclonal | |
Western Blot | |
widely interspaced zinc finger motifs, widely-interspaced zinc finger motifs, Widely-interspaced zinc finger-containing protein, WIZ zinc finger, Zinc finger protein 803, ZNF803protein Wiz | |
Recognizes region between residues Between between 1601 and 1651.. | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
58525 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction