Learn More
Moore Medical Wizard Wand Eye Magnet Light Kit
Comprehensive, portable kit for the efficient, professional removal of particles in the eye
Manufacturer:  Moore Medical 58928
Catalog No. 19-815-396
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes:
  • Comes with pocket clip and 2 AAA batteries
  • Lighted nylon eye loop attachment
  • Lighted eye magnet attachment
  • Spare lamp module with 4000 candlepower xenon bulb
  • Fluorescein strips
Description
Description
Innovative and lightweight, the Wizard Wand facilitates the examination and removal of foreign bodies and/or corneal abrasions in the eye
  • Collimated, diffused, narrow spot of light prevents shadows and irregularities in the illuminated area
  • Cobalt blue light excites fluorescein dyes to locate corneal abrasions and translucent particles
  • Use eye loop and eye magnet attachments to remove particles
  • Clean kit components with any cold sterilization solution such as Cidex, alcohol or peroxide.
Specifications
Specifications
Facilitates the examination of eyes for foreign bodies and/or corneal abrasions
Cobalt blue 4000 candlepower
Each
2 AAA batteries, lamp replacement, fluorescein strips, nylon loop tip and magnet tip
Wizard Wand Eye Care Kit
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Do not attempt to remove embedded particles with lighted eye magnet.