Learn More
Thermo Scientific™ WKITS™ Water QC Kit 1003
Thermo Scientific™ WKIT Water QC Kit 1003 is used for water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.
Supplier:  Thermo Scientific™ R4741003
Catalog No. R4741003
Add to Cart
This item is not returnable. View return policy
Description
Description
The Thermo Scientific™ WKIT Water QC Kit 1003 is used for water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.
  • WaterBugs™ deliver <50 CFUs in a single inoculum to mimic water samples
Ordering Alerts:
Reagents are sufficient for one test/reconstituted vial.
Includes:
Contents: 10 preserved organism vials (5 per organism); 10 rehydration vials. Escherichia coli strain C1, Pseudomonas aeruginosa ATCC™ 27853™†
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Water QC Kit 1003
Quality Control Kit
2/Pk.
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.