Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ WNK2 Recombinant Human Protein
WNK2 is a serine⁄threonine kinase involved in signal transduction and cellular communication
Supplier: Thermo Scientific™ PV4341

Description
- A serine/threonine kinase involved in signal transduction and cellular communication
Specifications
NP_006639.3 | |
64.9 kDa | |
WNK2 | |
aa 166-489 | |
GST Tag | |
Biochemical binding/kinetic | |
Dry Ice | |
WNK lysine deficient protein kinase 2 | |
Kinases & Inhibitors | |
Liquid | |
Kinase Assay | |
Signal Transduction |
65268 | |
10 µg | |
WNK Kinase Family | |
Yes | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: Other | |
WNK2 | |
Baculovirus | |
Recombinant, Catalytic | |
Serine⁄Threonine Kinases | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title