Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ WNK2 Recombinant Human Protein

WNK2 is a serine⁄threonine kinase involved in signal transduction and cellular communication

$451.00

Specifications

Accession Number NP_006639.3
Expression System Baculovirus
Molecular Weight 64.9 kDa
Protein Tag GST Tag
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV4341 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific™
PV4341
10 µg
Each for $451.00
Add to cart
 
Description

Description

Specifications

Specifications

NP_006639.3
Baculovirus
64.9 kDa
GST Tag
Safety and Handling

Safety and Handling

SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit