Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Wnt-3a Protein
SDP

Highly purified. Generating reliable and reproducible results.

$398.00 - $1110.00

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol
Gene ID (Entrez) 89780
Molecular Weight (g/mol) TMW: 37.5kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25194601
SDP
View Documents
Novus Biologicals™
NBP2519460.1MG
0.1mg
Each for $398.00
Add to Cart
 
NBP25194605
SDP
View Documents
Novus Biologicals™
NBP2519460.5MG
0.5mg
Each for $1,110.00
Add to Cart
 
Description

Description

An un-tagged denatured recombinant protein corresponding to the amino acids 19-352 of Human Wnt-3a The Recombinant Human Wnt-3a Protein is derived from E. coli. The Recombinant Human Wnt-3a Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

0.5mg/mL
20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol
TMW: 37.5kDa
MGC119418, MGC119419, MGC119420, protein Wnt-3a, wingless-type MMTV integration site family, member 3A
Unconjugated
>85%, by SDS-PAGE
SDS-PAGE
89780
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Wnt-3a
Recombinant
SDS
Documents

Documents

Product Certifications

For research use only and is not approved for use in humans or in clinical diagnosis

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit