Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Wnt-3a Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB9025100
This item is not returnable.
View return policy
Description
Wnt-3a Monoclonal specifically detects Wnt-3a in Human, Mouse samples. It is validated for Neutralization.Specifications
Wnt-3a | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MGC119418, MGC119419, MGC119420, protein Wnt-3a, wingless-type MMTV integration site family, member 3A, Wnt3a | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Wnt-3a Ser19-Lys352 Accession # P27467 | |
100 μg | |
Primary | |
Detects human and mouse Wnt-3a in direct ELISAs. | |
Human, Mouse | |
Purified |
Neutralization | |
930769 | |
Neutralization 0.15-0.9 ug/mL | |
P27467 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
89780 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction