Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Wnt-4 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 6 publications
Supplier: R&D Systems AF475SP
This item is not returnable.
View return policy
Description
Wnt-4 Polyclonal specifically detects Wnt-4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Wnt-4 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
protein Wnt-4, SERKAL, wingless-type MMTV integration site family, member 4, Wnt4, WNT-4 | |
Goat | |
Affinity Purified | |
RUO | |
54361 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
P22724 | |
WNT4 | |
E. coli-derived recombinant mouse Wnt-4 Ile37-Glu76, Arg222-Thr295 Accession # P22724 | |
25 μg | |
Primary | |
Detects mouse Wnt-4 in direct ELISAs and Western blots. The antibody has also been shown to recognize a mammalian cell-expressed mouse Wnt-4 protein in Western blots. | |
Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction