Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wnt-5a Mouse anti-Human, Alexa Fluor™ 750, Clone: 4M5E4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP227295AF750

Catalog No. NP22729A70


Add to cart

Description

Description

Wnt-5a Monoclonal antibody specifically detects Wnt-5a in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Wnt-5a
4M5E4
Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
WNT5A
A portion of the human Wnt5A protein was used as the immunogen for this antibody.
Protein G purified
Cancer Stem Cells, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary
7474.0
Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
hWNT5A, protein Wnt-5a, wingless-type MMTV integration site family, member 5A, WNT-5A protein
Mouse
IgG2a Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit