Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wnt-5a Rabbit anti-Human, Mouse, Rat, Bovine, Monkey, Alexa Fluor™ 750, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP224752AF750

Catalog No. NP22472A70


Add to cart

Description

Description

Wnt-5a Polyclonal antibody specifically detects Wnt-5a in Human, Mouse, Rat, Bovine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Wnt-5a
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
hWNT5A, protein Wnt-5a, wingless-type MMTV integration site family, member 5A, WNT-5A protein
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Bovine, Human, Monkey, Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Affinity Purified, Conjugated
WNT5A
A synthetic peptide corresponding to amino acids 190-230 of human Wnt-5a was used as the immunogen for this antibody, GenBank no NP_003383.2.
Affinity purified
Cancer Stem Cells, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary
7474.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit