Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wnt-5b Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $320.71

Specifications

Antigen Wnt-5b
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Applications Western Blot, Immunohistochemistry
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125990
View Documents
Novus Biologicals
NBP31054425UL
25 μg
Each for $106.19
Add to cart
 
NB125989
View Documents
Novus Biologicals
NBP310544100UL
100 μg
Each for $320.71
Add to cart
 
Description

Description

Wnt-5b Polyclonal antibody specifically detects Wnt-5b in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

Wnt-5b
Western Blot, Immunohistochemistry
Purified
RUO
Human
MGC2648, protein Wnt-5b, wingless-type MMTV integration site family, member 5B, WNT-5B protein
The immunogen is a synthetic peptide directed towards the middle region of human Wnt-5b (NP_110402). Peptide sequence YCLRNESTGSLGTQGRLCNKTSEGMDGCELMCCGRGYNQFKSVQVERCHC
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml, Immunohistochemistry
Unconjugated
Rabbit
Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
PBS buffer, 2% sucrose
81029
Polyclonal
Affinity purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit