Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wnt-7a Mouse anti-Human, Clone: 946744, R&D Systems™

Mouse MonoclonalAntibody

$116.82 - $345.50

Specifications

Antigen Wnt-7a
Clone 946744
Dilution Immunocytochemistry 8-25 ug/mL
Applications Immunocytochemistry
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
MAB30081100 View Documents R&D Systems
MAB30081100
100 ug Unconjugated Each for $345.50
Add to cart
 
MAB30081SP View Documents R&D Systems
MAB30081SP
25 ug Unconjugated Each for $116.82
Add to cart
 
Description

Description

Wnt-7a Monoclonal antibody specifically detects Wnt-7a in Human samples. It is validated for Immunocytochemistry.
Specifications

Specifications

Wnt-7a
Immunocytochemistry 8-25 ug/mL
Purified
Human
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Chinese hamster ovary cell line CHO-derived recombinant human Wnt-7a Met1-Lys349 Accession # O00755
Monoclonal
Protein A or G purified from hybridoma culture supernatant
Detects human Wnt-7a in direct ELISAs.
946744
Immunocytochemistry
Mouse
RUO
7476
IgG1
Primary
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit