Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WNT10B Recombinant Rabbit Monoclonal Antibody (HL2370)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550508
Description
WNT10B Recombinant Monoclonal Antibody for Western Blot, IHC (P)
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It may be involved in breast cancer, and its protein signaling is likely a molecular switch that governs adipogenesis. This protein is 96% identical to the mouse Wnt10b protein at the amino acid level. This gene is clustered with another family member, WNT1, in the chromosome 12q13 region.
Specifications
WNT10B | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
O00744, P48614 | |
WNT10B | |
Recombinant fragment of human Wnt10b. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
HL2370 | |
Unconjugated | |
WNT10B | |
Protein Wnt-10b; Protein Wnt-12; SHFM6; wingless related MMTV integration site 10b; wingless-type MMTV integration site family member 10B; wingless-type MMTV integration site family, member 10B; Wnt family member 10B; Wnt10; WNT10B; Wnt-10b; WNT-10B protein; WNT12; wnt-12 | |
Rabbit | |
Protein A | |
RUO | |
22410, 315294, 7480 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction