Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wnt16 Antibody (HL1498) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325751
Description
Wnt16 Monoclonal antibody specifically detects Wnt16 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Wnt16 | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
HL1498 | |
Western Blot 1:500-1:3000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
protein Wnt-16, wingless-type MMTV integration site family, member 16 | |
Recombinant protein encompassing a sequence within the center region of human Wnt16. The exact sequence is proprietary. | |
100 μL | |
Wnt Signaling Pathway | |
51384 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction