Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wnt3 Mouse anti-Human, DyLight™ 488, Clone: OTI1G7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274869G

Catalog No. NBP274869G


Add to cart

Description

Description

Wnt3 Monoclonal antibody specifically detects Wnt3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Wnt3
OTI1G7
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
WNT3
Recombinant protein fragment corresponding to amino acids 128-355 of human Wnt3 (NP_110380) produced in E.coli.
Immunogen affinity purified
Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary
7473.0
Immunohistochemistry, Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
INT4Proto-oncogene Int-4 homolog, MGC131950, MGC138321, MGC138323, proto-oncogene Wnt-3, wingless-type MMTV integration site family, member 3, WNT-3 proto-oncogene protein
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit