Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wnt3 Antibody (OTI1G7), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274869F
Description
Wnt3 Monoclonal specifically detects Wnt3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
Wnt3 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
WNT3 | |
Recombinant protein fragment corresponding to amino acids 128-355 of human Wnt3 (NP_110380) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Immunohistochemistry | |
OTI1G7 | |
Immunohistochemistry | |
INT4Proto-oncogene Int-4 homolog, MGC131950, MGC138321, MGC138323, proto-oncogene Wnt-3, wingless-type MMTV integration site family, member 3, WNT-3 proto-oncogene protein | |
Mouse | |
Affinity Purified | |
Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
7473.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction