Learn More
WNT6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 242011AP
Catalog No. 50-173-4260
Add to cart
Description
Description
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It is overexpressed in cervical cancer cell line and strongly coexpressed with another family member, WNT10A, in colorectal cancer cell line. The gene overexpression may play key roles in carcinogenesis. This gene and the WNT10A gene are clustered in the chromosome 2q35 region. The protein encoded by this gene is 97% identical to the mouse Wnt6 protein at the amino acid level.
Specifications
Specifications
WNT6
0.51 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P22727, Q9Y6F9
Protein Wnt 6, WNT6
Rabbit
IgG
150 μL
-20°C
Primary
22420, 7475
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
WNT6
Liquid
WNT6
WNT6 Fusion Protein Ag20889
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse