Learn More
WNT7B Recombinant Protein, Abnova
Human WNT7B full-length ORF recombinant protein with GST-tag at N-terminal
Specifications
Cross Reactivity | Human |
---|---|
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blotting (Recombinant Protein) |
Molecular Weight | 60.72 |
Name | Human Wnt7b Full-length ORF Recombinant Protein with GST-tag at N-terminal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-011-080
|
Abnova Corporation
H00007477P0110 |
10μg |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
89-011-081
|
Abnova Corporation
H00007477P0125 |
25μg |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
This gene is a member of the WNT gene family, which consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. Among members of the human WNT family, this gene product is most similar to WNT7A protein.
- Molecular Weight: 60.72kDa
- Preparation Method: in vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 Fast Flow
- Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH 8.0 in the elution buffer
Sequence: LGANIICNKIPGLAPRQRAICQSRPDAIIVIGEGAQMGINECQYQFRFGRWNCSALGEKTVFGQELRVGSREAAFTYAITAAGVAHAVTAACSQGNLSNCGCDREKQGYYNQAEGWKWGGCSADVRYGIDFSRRFVDAREIKKNARRLMNLHNNEAGKKVLEDRMQLECKCHGVSGSCTTKTC
WTTLPKFREVGHLLEEKYNAAVQVEVVRASRLRQPTFLRIKQLRSYQKPMETDLVYIEKSPNYCEEDAATGSVGTQGRLCNRTSPGADGCDTMCCGRGYNTHQYTKVWQCNCKFHWCCFVKCNTCSERTEVFTCK
Best when used within three months from the date of receipt.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Human | |
Antibody Production, ELISA, Protein Array, Western Blotting (Recombinant Protein) | |
Human Wnt7b Full-length ORF Recombinant Protein with GST-tag at N-terminal | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE stained with Coomassie Blue | |
Wheat Germ (in vitro) |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
60.72 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Yes | |
-80°C |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.