Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WNVM, Rabbit anti-Human,Mouse,Hamster,Monkey,Virus, Biotin, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB10056743B

Catalog No. NB003741


Add to cart

Description

Description

WNVM Polyclonal specifically detects WNVM in Human, Mouse, Hamster, Monkey, Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

WNVM
Biotin
PBS with 0.05% Sodium Azide
West Nile Virus envelope glycoprotein M
A synthetic peptide corresponding to amino acid residues 8-27 (GESTLANKKGAWLDSTKATR) of West Nile Virus glycoprotein M. Genbank accession no. NP_776013.
Affinity purified
RUO
Primary
Human, Mouse, Hamster, Monkey, Virus
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Affinity Purified, Conjugated
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit